Learn More
3-(4-(N-Biotinoyl-6-aminocaproyloxy)phenyl)propionic Acid, N-Hydroxysuccinimide Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(4-(N-Biotinoyl-6-aminocaproyloxy)phenyl)propionic Acid, N-Hydroxysuccinimide Ester |
| CAS | 1322623-44-8 |
| Molecular Formula | C28H36N4O8S |
| Quantity | 10 mg |
| Synonym | BPE |
| InChI Formula | InChI=1S/C28H36N4O8S/c33-22(7-4-3-6-21-27-20(17-41-21)30-28(38)31-27)29-15-5-1-2-8-25(36)39-19-11-9-18(10-12-19)16-26(37)40-32-23(34)13-14-24(32)35/h9-12,20-21,27H,1-8,13-17H2,(H,29,33)(H2,30,31,38)/t20?,21-,27?/m0/s1 |
| SMILES | O=C(CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)NCCCCCC(=O)Oc3ccc(CC(=O)ON4C(=O)CCC4=O)cc3 |
| IUPAC Name | [4-[2-(2,5-dioxopyrrolidin-1-yl)oxy-2-oxoethyl]phenyl] 6-[5-[(4S)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]hexanoate |
| Molecular Weight (g/mol) | 588.67 |