Learn More
3,4-Dihydro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1(2H)-isoquinolinone, TRC
Specifications
Specifications
| Chemical Name or Material | 3,4-Dihydro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1(2H)-isoquinolinone |
| CAS | 376584-30-4 |
| Molecular Formula | C15H20BNO3 |
| Quantity | 500 mg |
| Synonym | 2-[1-Oxo-1,2,3,4-tetrahydro-6-isoquinolinyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydroisoquinolin-1(2H)-one |
| InChI Formula | InChI=1S/C15H20BNO3/c1-14(2)15(3,4)20-16(19-14)11-5-6-12-10(9-11)7-8-17-13(12)18/h5-6,9H,7-8H2,1-4H3,(H,17,18) |
| SMILES | CC1(C)C(C)(C)OB(C2=CC=C3C(=C2)CCNC3=O)O1 |
| Molecular Weight (g/mol) | 273.14 |
| Formula Weight | 273.1536 |