Learn More
3,4-Difluorobenzoyl chloride, 98%
Product Code. 11378025
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11378025
Brand: Thermo Scientific Alfa Aesar L08188.06
Request Bulk or Custom Format
CAS: 76903-88-3 | C7H3ClF2O | 176.55 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 76903-88-3
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.55
MDL Number MFCD00009930
InChI Key RPQWXGVZELKOEU-UHFFFAOYSA-N
Synonym 3,4-difluorobenzoylchloride, benzoyl chloride, 3,4-difluoro, 3,4-difluoro benzoyl chloride, pubchem14796, acmc-209p6r, 3,4 difluorobenzoylchloride, 3,4-diflorobenzoyl chloride, 3,4-difluorobezoyl chloride, 3,4-diflourobenzoyl chloride, ksc494e2f
PubChem CID 2724519
IUPAC Name 3,4-difluorobenzoyl chloride
SMILES FC1=CC=C(C=C1F)C(Cl)=O
Specifications
Density 1.402
Boiling Point 185°C to 186°C
Flash Point 78°C (172°F)
Refractive Index 1.511
Quantity 5 g
UN Number UN3265
Beilstein 4177619
Sensitivity Moisture sensitive
Formula Weight 176.55
Percent Purity 98%
Chemical Name or Material 3,4-Difluorobenzoyl chloride