Learn More
3,4-Difluoro-2-methylbenzoic acid, 97+%, Thermo Scientific™
Product Code. p-7082732
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15495037
Brand: Thermo Scientific Alfa Aesar H32326.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3,4-Difluoro-2-methylbenzoic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in medicine field, electronic industry and polymer material.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 157652-31-8
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.13
MDL Number MFCD07777153
InChI Key AGUUCAUQWFAFPR-UHFFFAOYSA-N
PubChem CID 12137678
IUPAC Name 3,4-difluoro-2-methylbenzoic acid
SMILES CC1=C(C=CC(F)=C1F)C(O)=O
Specifications
Melting Point 152°C to 156°C
Color White to Cream
Odor Odorless
Quantity 1 g
Synonym 3,4-difluoro-2-methylbenzoicacid, 3,4-difluoro-2-methyl-benzoic acid, 3,4-difluoro-o-toluic acid, benzoic acid, 3,4-difluoro-2-methyl, pubchem8240, benzoic acid,3,4-difluoro-2-methyl
Solubility Information Insoluble in water.
Formula Weight 172.13
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 3,4-Difluoro-2-methylbenzoic acid