Learn More
3-(4-Chlorophenoxy)pentane-2,4-dione, Tech., Thermo Scientific™
Product Code. 10427062
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10427062 1 g 1g
10263472 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10427062 Supplier Thermo Scientific Maybridge Supplier No. SPB03987DA
 Restricted Item
Estimated Shipment: 06-11-2026
Chemical Identifiers
CAS 31168-10-2
Molecular Formula C11H11ClO3
Molecular Weight (g/mol) 226.656
MDL Number MFCD00204239
InChI Key QOOLFYCEZZCVSE-UHFFFAOYSA-N
Synonym 3-4-chlorophenoxy pentane-2,4-dione, 2,4-pentanedione,3-4-chlorophenoxy, 3-4-chlorophenoxy-2,4-pentanedione
PubChem CID 2799447
IUPAC Name 3-(4-chlorophenoxy)pentane-2,4-dione
SMILES CC(=O)C(C(=O)C)OC1=CC=C(C=C1)Cl
Specifications
Quantity 1 g
Linear Formula ClC6H4OCHC(O)CH32
Formula Weight 226.66
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-(4-Chlorophenoxy)pentane-2, 4-dione