Learn More
3,4-Bis(dibromomethyl)benzonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 3,4-Bis(dibromomethyl)benzonitrile |
| CAS | 19047-19-9 |
| Molecular Formula | C23 H33 O4 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C23H33O4/c1-14(24)26-17-6-8-18-16(13-17)5-7-20-19(18)9-11-22(3)21(20)10-12-23(22,4)27-15(2)25/h5,13,17-21H,6-12H2,1-4H3/t18-,19-,20+,21+,22-,23?/m0/s1 |
| SMILES | CC(=O)OC1(C)CC[C@@H]2[C@@H]3CC=C4C=[CH](CC[C@@H]4[C@@H]3CC[C@]12C)OC(=O)C |
| IUPAC Name | [(8R,9R,10R,13S,14R)-17-acetyloxy-13,17-dimethyl-1,2,3,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-yl] acetate |
| Molecular Weight (g/mol) | 373.506 |
| Formula Weight | 373.238 |