Learn More
3-(4-Biphenyl)propionic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(4-Biphenyl)propionic Acid |
| CAS | 35888-99-4 |
| Molecular Formula | C24 H34 N4 O5 S |
| Quantity | 1 g |
| Synonym | 3-(Biphenyl-4-yl)propanoic Acid, 3-(p-Biphenyl)propionic Acid, 4-Biphenylpropionic Acid, p-Phenylhydrocinnamic Acid |
| InChI Formula | InChI=1S/C24H34N4O5S/c1-4-20-17(3)15-28(22(20)29)24(31)25-14-13-18-7-5-6-8-21(18)34(32,33)27-23(30)26-19-11-9-16(2)10-12-19/h5-8,16,19H,4,9-15H2,1-3H3,(H,25,31)(H2,26,27,30) |
| SMILES | CCC1=C(C)CN(C(=O)NCCc2ccccc2S(=O)(=O)NC(=O)NC3CCC(C)CC3)C1=O |
| IUPAC Name | 4-ethyl-3-methyl-N-[2-[2-[(4-methylcyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]-5-oxo-2H-pyrrole-1-carboxamide |
| Molecular Weight (g/mol) | 490.616 |
| Formula Weight | 490.225 |