Learn More
3,4,6-Tri-O-methyl-D-glucal, 97%
Product Code. 15477178
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15477178 250 mg 250mg
15467178 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15477178 Supplier Thermo Scientific Alfa Aesar Supplier No. H60694.MD
Request Bulk or Custom Format
CAS: 16740-98-0 | C9H16O4 | 188.22 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16740-98-0
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD22989010
InChI Key PZJPNKKRMNFDQJ-UHFFFAOYNA-N
Synonym tri-o-methyl-d-glucal, 3,4,6-tri-o-methyl-d-glucal, tri-o-methyl glucal, 2r,3s,4r-3,4-dimethoxy-2-methoxymethyl-3,4-dihydro-2h-pyran, 3-o,4-o,6-o-trimethyl-1,2-dideoxy-d-arabino-1-hexenopyranose
PubChem CID 11252452
SMILES COCC1OC=CC(OC)C1OC
Specifications
Quantity 250 mg
IUPAC Name 3,4-dimethoxy-2-(methoxymethyl)-3,4-dihydro-2H-pyran
Formula Weight 188.22
Percent Purity 97%
Chemical Name or Material 3,4,6-Tri-O-methyl-D-glucal