Learn More
3,4,5-Trihydroxybenzoic-2,6-d2 Acid, CDN
Product Code. 30321473
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321473 0.25 g 0.25g
30312343 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321473 Supplier CDN Supplier No. CDND27280.25G
 Restricted Item
Estimated Shipment: 09-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Gallic acid 2,6 d2
CAS 294660-92-7
Molecular Formula C7 2H2 H4 O5
Quantity 0.25 g
Synonym Gallic Acid D2 (2,6 D2), Benzoic-2,6-d2 acid, 3,4,5-trihydroxy- (9 CI)
InChI Formula InChI=1 S/C7H6O5/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1-2,8-10 H,(H,11,12)/i1D,2 D
SMILES [2 H]c1c(O)c(O)c(O)c([2 H])c1C(=O)O
IUPAC Name 2,6-dideuterio-3,4,5-trihydroxybenzoic acid
Molecular Weight (g/mol) 172.13
Formula Weight 172.0341 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity