Learn More
3,4,5-Trichloropyridine, 98%, Thermo Scientific™
Product Code. 15432242 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 15432242
Brand: Thermo Scientific Acros 458210250
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Chemical Identifiers
CAS 33216-52-3
Molecular Formula C5H2Cl3N
Molecular Weight (g/mol) 182.44
InChI Key KKWRVUBDCJQHBZ-UHFFFAOYSA-N
Synonym pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1g
PubChem CID 3786531
IUPAC Name 3,4,5-trichloropyridine
SMILES C1=C(C(=C(C=N1)Cl)Cl)Cl
Specifications
Melting Point 72.0°C to 73.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Formula Weight 182.44
Percent Purity 98%
Chemical Name or Material 3,4,5-Trichloropyridine