Learn More
3,4,5-Trichlorocatechol, TRC
Product Code. 30456915
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456915 500 mg 500mg
30452056 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30456915 Supplier Toronto Research Chemicals Supplier No. TRCT773925500MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56961-20-7
Molecular Formula C6 H3 Cl3 O2
Molecular Weight (g/mol) 213.45
Synonym 1,2-Benzenediol, 3,4,5-trichloro-, 3,4,5-Trichloro-1,2-benzenediol, 3,4,5-Trichloro-2-hydroxyphenol, 3,4,5-Trichlorocatechol, 3,4,5-Trichloropyrocatechol
IUPAC Name 3,4,5-trichlorobenzene-1,2-diol
SMILES Oc1cc(Cl)c(Cl)c(Cl)c1O
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C6H3Cl3O2/c7-2-1-3(10)6(11)5(9)4(2)8/h1,10-11H
Formula Weight 211.9199
Chemical Name or Material 3,4,5-Trichlorocatechol