missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diethylthiacarbocyanine iodide, 96%

Product Code. 15487187
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487187 250 mg 250mg
15477187 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487187 Supplier Thermo Scientific Alfa Aesar Supplier No. H34251.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 905-97-5 | C21H22IN2S2+ | 493.445 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 905-97-5
Molecular Formula C21H22IN2S2+
Molecular Weight (g/mol) 493.445
MDL Number MFCD00011956
InChI Key VZBILKJHDPEENF-UHFFFAOYSA-N
Synonym 3,3'-diethylthiacarbocyanine iodide
PubChem CID 129893628
IUPAC Name (2Z)-3-ethyl-2-[(E)-3-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzothiazole;hydroiodide
SMILES CCN1C2=CC=CC=C2SC1=CC=CC3=[N+](C4=CC=CC=C4S3)CC.I

Specifications

Melting Point ∼250°C (decomposition)
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 492.45
Percent Purity 96%
Chemical Name or Material 3,3'-Diethylthiacarbocyanine iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.