Learn More
3,3',5,5'-Tetramethoxy-bibenzyl, TRC
Specifications
Specifications
| Chemical Name or Material | 3,3',5,5'-Tetramethoxy-bibenzyl |
| CAS | 22976-41-6 |
| Molecular Formula | C18H22O4 |
| Quantity | 25 mg |
| Synonym | 1,1'-(1,2-Ethanediyl)bis[3,5-dimethoxy-benzene |
| InChI Formula | InChI=1S/C18H22O4/c1-19-15-7-13(8-16(11-15)20-2)5-6-14-9-17(21-3)12-18(10-14)22-4/h7-12H,5-6H2,1-4H3 |
| SMILES | COC1=CC(=CC(=C1)CCC2=CC(=CC(=C2)OC)OC)OC |
| IUPAC Name | 1-[2-(3,5-dimethoxyphenyl)ethyl]-3,5-dimethoxybenzene |
| Molecular Weight (g/mol) | 302.36 |
| Formula Weight | 302.1518 |