Learn More
(3,3,4,4,4-d5)-(E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-[4-(t-butyldimethylsilyloxy)phenyl]-2-phenyl-1-butene, TRC
Product Code. 30351926
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30351926 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30351926 Supplier Toronto Research Chemicals Supplier No. TRCC36575225MG
 Restricted Item
Estimated Shipment: 14-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (3,3,4,4,4-d5)-(E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-[4-(t-butyldimethylsilyloxy)phenyl]-2-phenyl-1-butene
CAS 1020719-26-9
Molecular Formula C30H32D5ClO2Si
Quantity 25 mg
Synonym 1-(2-Chloroethoxy)-4-[1-[4-[[(1,1-dimethylethyl)dimethylsilyl]oxy]phenyl]-2-phenyl-1-buten-1-yl-3,3,4,4,4-d5]benzene
InChI Formula InChI=1S/C30H37ClO2Si/c1-7-28(23-11-9-8-10-12-23)29(24-13-17-26(18-14-24)32-22-21-31)25-15-19-27(20-16-25)33-34(5,6)30(2,3)4/h8-20H,7,21-22H2,1-6H3/b29-28+/i1D3,7D2
SMILES [2H]C([2H])([2H])C([2H])([2H])\C(=C(\c1ccc(OCCCl)cc1)/c2ccc(O[Si](C)(C)C(C)(C)C)cc2)\c3ccccc3
IUPAC Name tert-butyl-[4-[(E)-1-[4-(2-chloroethoxy)phenyl]-3,3,4,4,4-pentadeuterio-2-phenylbut-1-enyl]phenoxy]-dimethylsilane
Molecular Weight (g/mol) 498.18
Formula Weight 497.2565