missing translation for 'onlineSavingsMsg'
Learn More

(3,3,3-Trifluoropropyl)trichlorosilane, 97%, Thermo Scientific Chemicals

Product Code. 11339315
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11339315

Brand: Thermo Scientific Alfa Aesar L16670.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

(3,3,3-Trifluoropropyl)trichlorosilane is used for the production of fluoro silicones liquids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3,3,3-Trifluoropropyl)trichlorosilane is used for the production of fluoro silicones liquids.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 592-09-6
Molecular Formula C3H4Cl3F3Si
Molecular Weight (g/mol) 231.50
MDL Number MFCD00013602
InChI Key WEUBQNJHVBMUMD-UHFFFAOYSA-N
Synonym trichloro 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trichlorosilane, silane, trichloro 3,3,3-trifluoropropyl, 3,3,3-trifluoropropyltrichlorosilane, acmc-20ajez, cf3ch2ch2sicl3, silane, trichloro trifluoropropyl, 3,3,3-trifluropropyltrichlorosiline, trichloro 3,3,3-trifluoropropyl silane #
PubChem CID 68963
IUPAC Name trichloro(3,3,3-trifluoropropyl)silane
SMILES FC(F)(F)CC[Si](Cl)(Cl)Cl

Tekniske data

Boiling Point 113°C to 114°C
Quantity 25 g
Linear Formula CF3CH2CH2SiCl3
UN Number UN2985
Beilstein 1748876
Flash Point 15°C (59°F)
Solubility Information Reacts with water.
Formula Weight 231.51
Percent Purity 97%
Refractive Index 1.386
Sensitivity Moisture sensitive
Density 1.395
Chemical Name or Material (3,3,3-Trifluoropropyl)trichlorosilane
Vis mere Vis mindre

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.