missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Thienyl)acrylic Acid, TRC

Product Code. 30454014
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454014 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454014 Supplier Toronto Research Chemicals Supplier No. TRCT34437025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(2-Thienyl)acrylic Acid
CAS 1124-65-8
Molecular Formula C7H6O2S
Quantity 25 g
Synonym 3-(2-Thienyl)-2-propenoic Acid, 2-Thiopheneacrylic Acid, 2-Thienylacrylic Acid, 3-(2-Thienyl)propenoic Acid, 3-(Thien-2-yl)acrylic Acid, 3-(Thiophene-2-yl)acrylic Acid, 3-Thiophen-2-ylacrylic Acid, NSC 4247
InChI Formula InChI=1S/C7H6O2S/c8-7(9)4-3-6-2-1-5-10-6/h1-5H,(H,8,9)/b4-3+
SMILES OC(=O)\C=C\c1cccs1
IUPAC Name (E)-3-thiophen-2-ylprop-2-enoic acid
Molecular Weight (g/mol) 154.19
Formula Weight 154.0089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.