Learn More
3-(2-Hydroxypropan-2-yl)phenylboronic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(2-Hydroxypropan-2-yl)phenylboronic Acid |
| CAS | 955369-43-4 |
| Molecular Formula | C9H13BO3 |
| Quantity | 250 mg |
| Synonym | (3-(2-Hydroxypropan-2-yl)phenyl)boronic acid |
| InChI Formula | InChI=1S/C9H13BO3/c1-9(2,11)7-4-3-5-8(6-7)10(12)13/h3-6,11-13H,1-2H3 |
| SMILES | CC(C)(O)C1=CC=CC(B(O)O)=C1 |
| IUPAC Name | [3-(2-hydroxypropan-2-yl)phenyl]boronic acid |
| Molecular Weight (g/mol) | 180.01 |
| Formula Weight | 180.0958 |