Learn More
3-[2-(Diaminomethyleneamino)-1,3-thiazol-4-ylmethylsulphinyl]-N-sulphamoylpropanamide (Famotidine Metabolite), TRC
Product Code. 30357982
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357982 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30357982 Supplier Toronto Research Chemicals Supplier No. TRCD416367100MG
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material 3-[2-(Diaminomethyleneamino)-1,3-thiazol-4-ylmethylsulfinyl]-N-sulfamoylpropanamide
CAS 1020719-36-1
Purity Grade Notes HPLC
Molecular Formula C8 H14 N6 O4 S3
Quantity 100 mg
Synonym 3-[[[2-[(Diaminomethylene)-amino]thiazol-4-yl]methyl]sulfinyl]-N-sulfamoyl-propanamide, Famotidine Imp. I (Pharmeuropa)
InChI Formula InChI=1S/C8H14N6O4S3/c9-7(10)13-8-12-5(3-19-8)4-20(16)2-1-6(15)14-21(11,17)18/h3H,1-2,4H2,(H,14,15)(H2,11,17,18)(H4,9,10,12,13)
SMILES NC(=Nc1nc(CS(=O)CCC(=O)NS(=O)(=O)N)cs1)N
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfinyl]-N-sulfamoylpropanamide
Molecular Weight (g/mol) 354.43
Formula Weight 354.0239
Percent Purity >95
Mehr anzeigen Weniger anzeigen