missing translation for 'onlineSavingsMsg'
Learn More

3-(2,3-Dihydroxyphenyl)propionic Acid, TRC

Product Code. 30372758
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372758 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372758 Supplier Toronto Research Chemicals Supplier No. TRCD4523751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(2,3-Dihydroxyphenyl)propionic Acid
CAS 3714-73-6
Molecular Formula C9 H10 O4
Quantity 1 g
Synonym Benzenepropanoic acid, 2,3-dihydroxy-, Hydrocinnamic acid, 2,3-dihydroxy- (6CI,7CI,8CI), 2,3-Dihydroxybenzenepropanoic acid, 3-(2,3-Dihydroxyphenyl)propanoic acid, 3-(2,3-Dihydroxyphenyl)propionic acid, β-(2,3-Dihydroxyphenyl)propionic acid, 2,3-Dihydroxyhydrocinnamic acid
InChI Formula InChI=1S/C9H10O4/c10-7-3-1-2-6(9(7)13)4-5-8(11)12/h1-3,10,13H,4-5H2,(H,11,12)
SMILES OC(=O)CCc1cccc(O)c1O
IUPAC Name 3-(2,3-dihydroxyphenyl)propanoic acid
Molecular Weight (g/mol) 182.17
Formula Weight 182.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.