Learn More
3-(1H-Pyrazol-1-yl)alanine-d3, TRC
Product Code. 30412077
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412077 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30412077 Supplier Toronto Research Chemicals Supplier No. TRCP84224710MG
 Restricted Item
Estimated Shipment: 02-10-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-(1H-Pyrazol-1-yl)alanine-d3
CAS 2468771-07-3
Molecular Formula C6H6D3N3O2
Quantity 10 mg
InChI Formula InChI=1S/C6H9N3O2/c7-5(6(10)11)4-9-3-1-2-8-9/h1-3,5H,4,7H2,(H,10,11)/t5-/m0/s1/i1D,2D,3D
SMILES N[C@@H](CN1N=C([2H])C([2H])=C1[2H])C(O)=O
IUPAC Name (S)-2-amino-3-(1H-pyrazol-1-yl-d3)propanoic acid
Molecular Weight (g/mol) 158.17
Formula Weight 158.09