Learn More
Biochrom 2x Micro-Volume Cuvettes with 0.5 mm (1x) and 0.125 mm (1x) Pathlength for 15 mm Beam Height Spectrophotometers
Click to view available options
Type:
Microvolume Cuvette
Unit Size:
Each
Description
- Exceptional measurement accuracy
- Wide measurement range - no more changing pathlengths
- Use only 1 μL of preicous sample
- Simple to use and clean
- Easy to check for sample impurities such as bubbles or dust
- Compatible with most standatd UV-Visible Sprectrophotometers
Specifications
Specifications
| Description | 2 x Micro-Volume Cuvettes with 0.5 mm (1 x) and 0.125 mm (1 x) Pathlength for 15 mm Beam Height Spectrophotometers |
| Dimensions (L x W x H) | 12.5 x 12.5 x 61.0 mm |
| Type | Microvolume Cuvette |
| Disposable | Reusable |
| For Use With (Equipment) | 15 mm Beam Height Spectrophotometers |
| Pathlength | 0.125 and 0.5 mm |
| Shape | Rectangular |
For research use only.