Learn More
(2S,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 5505-63-5 |
| Molecular Formula | C6H14ClNO5 |
| MDL Number | MFCD09880213 |
| Quantity | 10 g |
| SMILES | O=C[C@@H](N)[C@@H](O)[C@H](O)[C@H](O)CO.[H]Cl |
| Molecular Weight (g/mol) | 215.63 |
| Percent Purity | 98% |
| Chemical Name or Material | (2S,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride |