Learn More
(2S,3R,4E)-3-Benzoyl-2-tert-butyloxycarbonylamino-4-octadecen-1,3-diol, TRC
Specifications
Specifications
| Chemical Name or Material | (2S,3R,4E)-3-Benzoyl-2-tert-butyloxycarbonylamino-4-octadecen-1,3-diol |
| CAS | 299172-59-1 |
| Molecular Formula | C30 H49 N O5 |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C30H49NO5/c1-5-6-7-8-9-10-11-12-13-14-15-16-20-23-27(35-28(33)25-21-18-17-19-22-25)26(24-32)31-29(34)36-30(2,3)4/h17-23,26-27,32H,5-16,24H2,1-4H3,(H,31,34)/b23-20+/t26-,27+/m0/s1 |
| SMILES | CCCCCCCCCCCCC\C=C\[C@@H](OC(=O)c1ccccc1)[C@H](CO)NC(=O)OC(C)(C)C |
| IUPAC Name | [(E,2S,3R)-1-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]octadec-4-en-3-yl] benzoate |
| Molecular Weight (g/mol) | 503.71 |
| Formula Weight | 503.3611 |