Learn More
(2S,3aS,7aS)-Octahydroindole-2-carboxylic acid, 98%
Description
An intermediate in the synthesis of Perindopril. The completely diastereoselective α-alkylation of oxazolidinone provides a very convenient and concise route to enantiopure α-tetrasubstituted derivatives of this Oic stereoisomer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
An intermediate in the synthesis of Perindopril. The completely diastereoselective α-alkylation of oxazolidinone provides a very convenient and concise route to enantiopure α-tetrasubstituted derivatives of this Oic stereoisomer.
Solubility
Soluble in methanol and water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at -20°C.
Specifications
Specifications
| Melting Point | 266°C (decomposition) |
| Quantity | 5 g |
| Solubility Information | Soluble in methanol and water. |
| Optical Rotation | −50 (c=1 in Methanol) |
| Formula Weight | 169.22 |
| Percent Purity | 98% |
| Chemical Name or Material | (2S,3aS,7aS)-Octahydroindole-2-carboxylic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.