Learn More
(2S,3aS,7aS)-1-Fmoc-octahydroindole-2-carboxylic acid, 98%
Product Code. 15458336
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
15458336 250 mg 250mg
15448336 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Product Code. 15458336 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' H63491.MD
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
CAS: 130309-37-4 | C24H25NO4 | 391.467 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 130309-37-4
Molecular Formula C24H25NO4
Molecular Weight (g/mol) 391.467
MDL Number MFCD00191201
InChI Key JBZXLQHJZHITMW-RXYZOABWSA-N
Synonym fmoc-oic-oh, 2s,3as,7as-1-9h-fluoren-9-yl methoxy carbonyl octahydro-1h-indole-2-carboxylic acid, fmoc-2s,3as,7as-octahydro-1h-indole-2-carboxylic acid, 2s,3as,7as-1-9h-fluoren-9-ylmethoxy carbonyl-octahydroindole-2-carboxylic acid, fmoc-l-octahydroindole-2-carboxylic acid, ambotzfaa1729, 2s pound not3as pound not7as-1-9h-fluoren-9-yl methoxy carbonyl, 2s,3as,7as-1-9h-fluoren-9-yl methoxy carbonyl-octahydro-1h-indole-2-carboxylic acid, 2s,3as,7as-1-9h-fluoren-9-ylmethoxycarbonyl-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid
PubChem CID 2756165
IUPAC Name (2S,3aS,7aS)-1-(9H-fluoren-9-ylmethoxycarbonyl)-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid
SMILES C1CCC2C(C1)CC(N2C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O
Specifications
Color White
Quantity 250 mg
Formula Weight 391.46
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (2S,3aS,7aS)-1-Fmoc-octahydroindole-2-carboxylic acid