Learn More
(2R,4S)-3-Benzoyl-4-methyl-2-phenyl-5-oxazolidinone, TRC
Specifications
Specifications
| Chemical Name or Material | (2R,4S)-3-Benzoyl-4-methyl-2-phenyl-5-oxazolidinone |
| CAS | 113806-28-3 |
| Molecular Formula | C17 H15 N O3 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C17H15NO3/c1-12-17(20)21-16(14-10-6-3-7-11-14)18(12)15(19)13-8-4-2-5-9-13/h2-12,16H,1H3/t12-,16+/m0/s1 |
| SMILES | C[C@@H]1N([C@H](OC1=O)c2ccccc2)C(=O)c3ccccc3 |
| IUPAC Name | (2R,4S)-3-benzoyl-4-methyl-2-phenyl-1,3-oxazolidin-5-one |
| Molecular Weight (g/mol) | 281.31 |
| Formula Weight | 281.1052 |