Learn More
(2R,3R,4R,5S)-2-(Hydroxymethyl)-1-[2-(phenylmethoxy)ethyl]-3,4,5-piperidinetriol, TRC
Specifications
Specifications
| Chemical Name or Material | (2R,3R,4R,5S)-2-(Hydroxymethyl)-1-[2-(phenylmethoxy)ethyl]-3,4,5-piperidinetriol |
| CAS | 1203659-54-4 |
| Molecular Formula | C15H23NO5 |
| Quantity | 2.5 mg |
| InChI Formula | InChI=1S/C15H23NO5/c17-9-12-14(19)15(20)13(18)8-16(12)6-7-21-10-11-4-2-1-3-5-11/h1-5,12-15,17-20H,6-10H2/t12-,13-,14+,15+/m0/s1 |
| SMILES | OC[C@H]1[C@@H](O)[C@H](O)[C@@H](O)CN1CCOCc2ccccc2 |
| IUPAC Name | (2S,3R,4R,5S)-2-(hydroxymethyl)-1-(2-phenylmethoxyethyl)piperidine-3,4,5-triol |
| Molecular Weight (g/mol) | 297.35 |
| Formula Weight | 297.1576 |