Learn More
Gilson™ 2nd Level Service Kit
Brand: Gilson™ F161978
This item is not returnable.
View return policy
Description
Specifications
| For Gilson Pipetman Classic P1000 Pipet | |
| Push button, connecting nut, tip ejector, button assembly, power tub cover, ejector window |