Learn More
(2E)-6-[[(1,1-Dimethylethoxy)carbonyl]amino]-2-hexenoic Acid Ethyl Ester, TRC
Product Code. 30006029
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006029 50 mg 50mg
30007490 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30006029 Supplier Toronto Research Chemicals Supplier No. TRCD47338050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1008751-73-2
Molecular Formula C13 H23 N O4
Molecular Weight (g/mol) 257.33
IUPAC Name ethyl (E)-6-(tert-butoxycarbonylamino)hex-2-enoate
SMILES CCOC(=O)\C=C\CCCNC(=O)OC(C)(C)C
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C13H23NO4/c1-5-17-11(15)9-7-6-8-10-14-12(16)18-13(2,3)4/h7,9H,5-6,8,10H2,1-4H3,(H,14,16)/b9-7+
Formula Weight 257.1627
Chemical Name or Material (2E)-6-[((1,1-Dimethylethoxy)carbonyl)amino]-2-hexenoic Acid Ethyl Ester