Learn More
(2E,4E)-2,4-Decadienoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (2E,4E)-2,4-Decadienoic Acid |
| CAS | 30361-33-2 |
| Molecular Formula | C10H16O2 |
| Quantity | 2.5 g |
| Synonym | (E,E)-2,4-Decadienoic Acid, 2(E),4(E)-Decadienoic Acid, 2-trans,4-trans-Decadienoic Acid, trans-2-trans-4-Decadienoic Acid |
| InChI Formula | InChI=1S/C10H16O2/c1-2-3-4-5-6-7-8-9-10(11)12/h6-9H,2-5H2,1H3,(H,11,12)/b7-6+,9-8+ |
| SMILES | CCCCC/C=C/C=C/C(=O)O |
| IUPAC Name | (2E,4E)-deca-2,4-dienoic acid |
| Molecular Weight (g/mol) | 168.23 |
| Formula Weight | 168.115 |