Learn More
Fisherbrand™ TV100YK-2DSYS TV Modular 2-D Vertical Electrophoresis System
Product Code. 12336517
Click to view available options
:
TV100YK-2DSYS TV Modular 2-D Vertical Electrophoresis System
Product Code. 12336517
Brand: Fisherbrand™ TV100YK2DSYS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Provides a complete solution within one gel tank for 2-D electrophoresis.
  • Comprises of one standard GRM and one CAPGRM, which are interchangeable within the same gel tank, allowing to perform PAGE and IEF independently or as part of a fully integrated 2-D system
  • CAPGRM is available in 2 sizes for first dimension electrophoresis of up to 10 ampholyte capillary gels, a maximum 8 or 17cm in length and 1 or 4mm in internal diameter
  • Capillary gels are available in standard 1mm internal diameter for analytical IEF, or as optional 4mm capillaries for protein isolation
  • Twin-plate standard GRM designed specifically for SDS-PAGE techniques requiring self-cast gels
  • Casting base is included as standard
  • Snaplock connectors allows the cooling coil to be connected to an external chiller unit for faster, higher voltage separations
Specifications
Type TV Modular 2-D Vertical Electrophoresis System
Includes Twin-plate mini-gel unit with GRM and cooled gel tank, with built-in cooling coil and quick-fit tubing, lid, 2 x (10 x 10cm; W x H) plain glass plates, 2 x (10 x 10cm) notched glass plates, 1 x dummy plate, 4 x 1mm thick spacers, 2 x spacer aligners and 2 x 1mm thick 12-sample combs, casting base and 2 x silicone seals; CAPGRM, 10 x 1mm glass capillaries, 4 x 1.5mm thick spacers, 2 x 1.5mm thick 1-D preparatory combs, 10 x blanking ports and 1 x gel extraction platform
Electrical Requirements 10 to 15 mA
Voltage 100 to 150 V