Learn More
2a-Cyano-ethisterone, TRC
Specifications
Specifications
| Chemical Name or Material | 2alpha-Cyano-ethisterone |
| CAS | 13648-12-9 |
| Molecular Formula | C22H27NO2 |
| Quantity | 250 mg |
| Synonym | (2S,8R,9S,10R,13S,14S,17R)-17-Ethynyl-17-hydroxy-10,13-dimethyl-3-oxo-2,3,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-2-carbonitrile, 17-Hydroxy-3-oxo-17alpha-Pregn-4-en-20-yne-2alpha-carbonitrile |
| InChI Formula | InChI=1S/C22H27NO2/c1-4-22(25)10-8-18-16-6-5-15-11-19(24)14(13-23)12-20(15,2)17(16)7-9-21(18,22)3/h1,11,14,16-18,25H,5-10,12H2,2-3H3/t14-,16+,17-,18-,20-,21-,22-/m0/s1 |
| SMILES | C[C@]12C[C@@H](C#N)C(=O)C=C1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@@]4(O)C#C |
| IUPAC Name | (2S,8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-2-carbonitrile |
| Molecular Weight (g/mol) | 337.46 |
| Formula Weight | 337.2042 |