Learn More
Roland Vetter Laborbedarf™ Color Coded Marking Tape, 25mm
Product Code. p-8010938
Click to view available options
Color:
Blue
Red
Unit Size:
55m
Product Code. 11720454
Brand: Roland Vetter Laborbedarf™ ST1004
Safe for use in autoclaves, freezers and ovens. Roland Vetter Laborbedarf™ Color-Coded Marking Tape can be written on with marker, ballpoint and felt-tip pen, and even pencil.
  • Resistant to water, oil and acid
  • Temperature -20 to + 121°C
  • For autoclave, freezer, refrigerator and oven
  • Marking with a ballpoint pen, pencil and felt-tip pen
Specifications
Color Red
Format Roll
Product Type Marking Tape
Size (Metric) 25 mm