Learn More
Verubecestat, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Verubecestat |
| CAS | 1286770-55-5 |
| Molecular Formula | C17H17F2N5O3S |
| Quantity | 25 mg |
| Synonym | N-[3-[(5R)-3-Amino-5,6-dihydro-2,5-dimethyl-1,1-dioxido-2H-1,2,4-thiadiazin-5-yl]-4-fluorophenyl]-5-fluoro-2-pyridinecarboxamide, MK 8931 |
| InChI Formula | InChI=1S/C17H17F2N5O3S/c1-17(9-28(26,27)24(2)16(20)23-17)12-7-11(4-5-13(12)19)22-15(25)14-6-3-10(18)8-21-14/h3-8H,9H2,1-2H3,(H2,20,23)(H,22,25)/t17-/m0/s1 |
| SMILES | CN1C(=N[C@@](C)(CS1(=O)=O)c2cc(NC(=O)c3ccc(F)cn3)ccc2F)N |
| IUPAC Name | N-[3-[(5R)-3-amino-2,5-dimethyl-1,1-dioxo-6H-1,2,4-thiadiazin-5-yl]-4-fluorophenyl]-5-fluoropyridine-2-carboxamide |
| Molecular Weight (g/mol) | 409.41 |
| Formula Weight | 409.102 |