missing translation for 'onlineSavingsMsg'
Learn More

Perindopril L-Arginine, TRC

Product Code. 18081650
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18081650 25 mg 25mg
18023850 1 mg 1mg
18083440 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18081650 Supplier Toronto Research Chemicals Supplier No. TRCP28760025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612548-45-5
Molecular Formula C25H46N6O7
Molecular Weight (g/mol) 542.67
Synonym L-Arginine, (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate (1:1), L-Arginine, mono[(2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate] (9CI), (2S)-2-Amino-5-guanidinopentanoic acid (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]propanoyl]octahydro-1H-indole-2-carboxylate, (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate L-Arginine (1:1), mono[(2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylate]
IUPAC Name (2S,3aS,7aS)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxopentan-2-yl]amino]propanoyl]-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid;(2S)-2-amino-5-carbamimidamidopentanoic acid
SMILES CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(=O)O)C(=O)OCC.N[C@@H](CCCNC(=N)N)C(=O)O

Specifications

InChI Formula InChI=1S/C19H32N2O5.C6H14N4O2/c1-4-8-14(19(25)26-5-2)20-12(3)17(22)21-15-10-7-6-9-13(15)11-16(21)18(23)24;7-4(5(11)12)2-1-3-10-6(8)9/h12-16,20H,4-11H2,1-3H3,(H,23,24);4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t12-,13-,14-,15-,16-;4-/m00/s1
Quantity 25 mg
Formula Weight 542.3428
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Perindopril L-Arginine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.