Learn More
Epsilon-Caprolactam-d10, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | eta-Caprolactam-d10 |
| CAS | 169297-53-4 |
| Molecular Formula | C6HD10NO |
| Quantity | 25 mg |
| Synonym | Hexahydro-3,4,5,6,7-d5-2H-azepin-2-one-3,4,5,6,7-d5 |
| InChI Formula | InChI=1S/C6H11NO/c8-6-4-2-1-3-5-7-6/h1-5H2,(H,7,8)/i1D2,2D2,3D2,4D2,5D2 |
| SMILES | [2H]C1([2H])NC(=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H] |
| IUPAC Name | 3,3,4,4,5,5,6,6,7,7-decadeuterioazepan-2-one |
| Molecular Weight (g/mol) | 123.22 |
| Formula Weight | 123.1468 |