missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Tryptophan-d5 (major), TRC
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
Specifications
Specifications
| Chemical Name or Material | DL-Tryptophan-d5 |
| Molecular Formula | C11H7D5N2O2 |
| Quantity | 25 mg |
| Synonym | (±)-Tryptophan-d5 (major), 1H-Indole-3-alanine-d5 (major), DL-α-Amino-3-indolepropionic Acid-d5 |
| InChI Formula | InChI=1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/i1D,2D,3D,4D,6D |
| SMILES | NC(C(O)=O)CC1=C([2H])NC2=C([2H])C([2H])=C([2H])C([2H])=C21 |
| Molecular Weight (g/mol) | 209.26 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction