missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bisphenol A-d6, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Bisphenol A-d6 |
| CAS | 86588-58-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 2H6 H10 O2 |
| Quantity | 25 mg |
| Synonym | Phenol, 4,4'-[1-(methyl-d3)ethylidene-2,2,2-d3]bis-, 4,4'-[1-(Methyl-d3)ethylidene-2,2,2-d3]bis[phenol], 2,2-Bis(4-hydroxyphenyl)-1,1,1,3,3,3-hexadeuteriopropane, 4,4'-Isopropylidenediphenol (isopropyl-D6), Bisphenol A (propane-D6), Bisphenol A (dimethyl-D6) |
| InChI Formula | InChI=1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])C(c1ccc(O)cc1)(c2ccc(O)cc2)C([2H])([2H])[2H] |
| IUPAC Name | 4-[1,1,1,3,3,3-hexadeuterio-2-(4-hydroxyphenyl)propan-2-yl]phenol |
| Molecular Weight (g/mol) | 234.32 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction