missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkynyl Myristic Acid (>90%), TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Alkynyl Myristic Acid |
| CAS | 82909-47-5 |
| Molecular Formula | C14 H24 O2 |
| Quantity | 25 mg |
| Synonym | 13-Tetradecynoic acid |
| InChI Formula | InChI=1S/C14H24O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h1H,3-13H2,(H,15,16) |
| SMILES | OC(=O)CCCCCCCCCCCC#C |
| IUPAC Name | tetradec-13-ynoic acid |
| Molecular Weight (g/mol) | 224.339 |
| Formula Weight | 224.178 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction