missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitromethane-d3 (Reagent grade), CDN
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
Specifications
Specifications
| Chemical Name or Material | Nitromethane-D3 |
| CAS | 13031-32-8 |
| Molecular Formula | C D3 N O2 |
| Quantity | 25 g |
| InChI Formula | InChI=1 S/CH3NO2/c1-2(3)4/h1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])[N+](=O)[O-] |
| IUPAC Name | trideuterio(nitro)methane |
| Molecular Weight (g/mol) | 64.058 |
| Formula Weight | 64.035 g/mol |
| Percent Purity | 99 atom % D, min 99% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction