Learn More
WST-1, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | WST-1 |
| CAS | 150849-52-8 |
| Molecular Formula | C19 H11 I N5 O8 S2 . Na |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C19H12IN5O8S2.Na/c20-12-1-3-13(4-2-12)23-21-19(22-24(23)14-5-7-15(8-6-14)25(26)27)17-10-9-16(34(28,29)30)11-18(17)35(31,32)33;/h1-11H,(H-,28,29,30,31,32,33);/q;+1/p-1 |
| SMILES | [Na+].[O-][N+](=O)c1ccc(cc1)n2nc(n[n+]2c3ccc(I)cc3)c4ccc(cc4S(=O)(=O)[O-])S(=O)(=O)[O-] |
| IUPAC Name | sodium;4-[2-(4-iodophenyl)-3-(4-nitrophenyl)tetrazol-2-ium-5-yl]benzene-1,3-disulfonate |
| Molecular Weight (g/mol) | 651.34 |
| Formula Weight | 650.8991 |