Learn More
(R)-2-((4-nitrophenethyl)amino)-1-phenylethanol Hydrochloride, TRC
Product Code. 17984645
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984645 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17984645 Supplier Toronto Research Chemicals Supplier No. TRCN502255250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 521284-21-9
Molecular Formula C16 H18 N2 O3 . H Cl
Quantity 250 mg
Synonym (alphaR)-[[[2-(4-nitrophenyl)ethyl]amino]methyl]-benzenemethanol Monohydrochloride, (R)-2-((4-nitrophenethyl)amino)-1-phenylethanol Hydrochloride, Imp. F (EP) as hydrochloride: (1R)-2-[[2-(4-Nitrophenyl)ethyl]amino]-1-phenylethan-1-ol Hydrochloride, Benzenemethanol, α-[[[2-(4-nitrophenyl)ethyl]amino]methyl]-, hydrochloride (1:1), (αR)- (ACI), Benzenemethanol, α-[[[2-(4-nitrophenyl)ethyl]amino]methyl]-, monohydrochloride, (αR)- (9CI), (R)-2-[[2-(4-Nitrophenyl)-ethyl]amino]-1-phenylethanol monohydrochloride, (R)-2-[[2-(4-Nitrophenyl)ethyl]amino]-1-phenylethanol hydrochloride
InChI Formula InChI=1S/C16H18N2O3.ClH/c19-16(14-4-2-1-3-5-14)12-17-11-10-13-6-8-15(9-7-13)18(20)21;/h1-9,16-17,19H,10-12H2;1H/t16-;/m0./s1
SMILES Cl.O[C@@H](CNCCc1ccc(cc1)[N+](=O)[O-])c2ccccc2
IUPAC Name (1R)-2-[2-(4-nitrophenyl)ethylamino]-1-phenylethanol;hydrochloride
Molecular Weight (g/mol) 322.787
Formula Weight 322.108
Chemical Name or Material (1R)-2-[(2-(4-Nitrophenyl)ethyl)amino]-1-phenylethan-1-ol Hydrochloride