Learn More
Melford NPG [4-Nitrophenyl-β-glucuronide
Product Code. 30018553
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 30018553
Brand: Melford N120300.25
Spectrophotometric substrate for detection of B-D-Glucuronidase.
Spectrophotometric substrate for detection of B-D-Glucuronidase.