Learn More
Bromophos-ethyl, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Bromophos-ethyl |
| CAS | 4824-78-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H12 Br Cl2 O3 P S |
| Quantity | 250 mg |
| Synonym | Bromophos A, Bromophos-ethyl, Ethyl bromophos, Filariol, Filariol 60, Nexagan, Nexagan G, O,O-Diethyl O-(2,5-dichloro-4-bromophenyl) thiophosphate, O-(4-Bromo-2,5-dichlorophenyl) O,O-diethyl phosphorothioate, OMS 659 |
| InChI Formula | InChI=1S/C10H12BrCl2O3PS/c1-3-14-17(18,15-4-2)16-10-6-8(12)7(11)5-9(10)13/h5-6H,3-4H2,1-2H3 |
| SMILES | CCOP(=S)(OCC)Oc1cc(Cl)c(Br)cc1Cl |
| IUPAC Name | (4-bromo-2,5-dichlorophenoxy)-diethoxy-sulfanylidene-λ^{5}-phosphane |
| Molecular Weight (g/mol) | 394.05 |
| Show More |