missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benz[e]acephenanthrylene, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Benzo[b]fluoranthene |
| CAS | 205-99-2 |
| Molecular Formula | C20 H12 |
| Quantity | 250 mg |
| Synonym | Benz[e]acephenanthrylene, 2,3-Benzfluoranthene, 3,4-Benz[e]acephenanthrylene, 3,4-Benzfluoranthene, 3,4-Benzofluoranthene, BbF, Benzo[b]fluoranthene, Benzo[e]fluoranthene, NSC 89265 |
| InChI Formula | InChI=1S/C20H12/c1-2-7-14-13(6-1)12-19-16-9-4-3-8-15(16)18-11-5-10-17(14)20(18)19/h1-12H |
| SMILES | c1ccc2c(c1)c3cccc4c5ccccc5cc2c34 |
| IUPAC Name | Benzo[b]fluoranthene |
| Molecular Weight (g/mol) | 252.31 |
| Formula Weight | 252.0939 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction