Learn More
Androstenedione, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Androstenedione |
| CAS | 63-05-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H26 O2 |
| Quantity | 250 mg |
| Synonym | 17-Ketotestosterone, 3,17-Dioxoandrost-4-ene, 4-Androstene-3,17-dione, Androstenedione, Fecundin, NSC 9563, SKF 2170, δ4-Androstene-3,17-dione |
| InChI Formula | InChI=1S/C19H26O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-16H,3-10H2,1-2H3/t14-,15-,16-,18-,19-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CCC2=O |
| IUPAC Name | (8R,9S,10R,13S,14S)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-3,17-dione |
| Molecular Weight (g/mol) | 286.41 |
| Show More |