missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[2-(2-Aminoethoxy)ethoxy]acetic Acid, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | [2-(2-Aminoethoxy)ethoxy]acetic Acid |
| CAS | 134978-97-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6H13NO4 |
| Quantity | 250 mg |
| Synonym | 2-[2-(2-Aminoethoxy)ethoxy]acetic Acid, 8-Amino-3,6-dioxaoctanoic AcidNSC 626945, [2-(2-Aminoethoxy)ethoxy]acetic Acid |
| SMILES | NCCOCCOCC(=O)O |
| Molecular Weight (g/mol) | 163.17 |
| Formula Weight | 163.0845 |
| Percent Purity | >95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction