Learn More
Mettler Toledo Online CONDUCTIVITY STANDARD, 12.88MS, 250ml
Brand:  Mettler Toledo Online 51350094
CONDUCTIVITY STANDARD, 12.88MS, 250ml
Product Code. 10259351
  • ONLINE EXCLUSIVE PRICE
    34.00€
    30.14€ / 250mL
    Save 3.86€
    11% Off