Learn More
BTX™ 25-Well Plate Seal
Product Code. 15731068
Click to view available options
:
25-Well Plate Seal
Product Code. 15731068
Brand: BTX™ 4500012
25-Well Plate Seal is a sealing film for multi-well electroporation plates.
BTX Multi-well Electroporation plate sealing films are used to:
  • Prevent well cross-contamination during electroporation
  • Mark off wells that have been used
  • Maintain sterility in unused wells
Specifications
Color Clear
Autoclavable Not Autoclavable
No. of Wells 24
Adhesive Yes
Pierceable Yes
Plate Compatibility BTX 25-well Electroporation Plates
Fit for Purpose Sterile