Learn More
BRAND™ 24-well Insert 2in1, PET-Membrane
Product Code. 16114957 Shop All Brand Products
Change view
Click to view available options
Pore Size:
0.4 μm
8 μm
No. per Pack:
4 x 12
48 x 1
Unit Size:
Pack of 48
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size No. per Pack unitSize
16114957 0.4 μm 4 x 12 Pack of 48
16104957 0.4 μm 48 x 1 Pack of 48
16124957 8 μm 48 x 1 Pack of 48
16134957 8 μm 4 x 12 Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16114957 Supplier BRAND™ Supplier No. 782711
The cellGrade™ plus surface of the membranes ensures excellent cell attachment and growth. The highly transparent membranes give you good visibilty of your cell cultures, and the high pore density contributes to good permeability.
  • Use Inserts in a hanging or standing position
  • For 24-well plates or culture dishes
  • Achieve uniform and controlled cell growth due to surface treatment, without coatings
TRUSTED_SUSTAINABILITY
Specifications
Description 24-well Insert 2in1, PET-Membrane 0.4 μm, Multipack
Material PET
Pore Size 0.4 μm
No. per Pack 4 x 12
Sterility Sterile
No. of Wells 24